Assay Details
| Gene Symbol: lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078877.1 | NP_001072345.1 | 9 - 10 | 1191 | 65 | |
| XM_012957259.2 | XP_012812713.1 | 8 - 9 | 843 | 65 | ||
| XM_018091339.1 | XP_017946828.1 | 7 - 8 | 975 | 65 | ||
| GenBank mRNA | BC121434.1 | - | 9 - 10 | 1191 | 65 | |
| BC135680.1 | - | 9 - 10 | 1184 | 65 | ||
More Information
Set Membership: |
|

lima1
Genomic Map