Assay Details
| Gene Symbol: tlr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078891.1 | NP_001072359.1 | 5 - 6 | 1142 | 99 | |
| GenBank mRNA | BC121458.1 | - | 5 - 6 | 1142 | 99 | |
More Information
Set Membership: |
|

tlr5
Genomic Map