Assay Details
Gene Symbol: il1rapl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078899.1 | NP_001072367.1 | 8 - 9 | 1612 | 63 | |
GenBank mRNA | BC121469.1 | - | 8 - 9 | 1612 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
