Assay Details
| Gene Symbol: acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078900.1 | NP_001072368.1 | 8 - 9 | 796 | 80 | |
| GenBank mRNA | BC121470.1 | - | 8 - 9 | 796 | 80 | |
| Gene Symbol: acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004916190.3 | XP_004916247.1 | 8 - 9 | 1658 | 80 | |
| XM_018095453.1 | XP_017950942.1 | 7 - 8 | 1641 | 80 | ||
More Information
Set Membership: |
|

acot7
Genomic Map