Assay Details
| Gene Symbol: cdipt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078906.1 | NP_001072374.1 | 3 - 4 | 343 | 74 | |
| GenBank mRNA | BC121479.1 | - | 3 - 4 | 343 | 74 | |
More Information
Set Membership: |
|

cdipt
Genomic Map