Assay Details
| Gene Symbol: ifnar2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078923.1 | NP_001072391.1 | 1 - 2 | 132 | 101 | |
| GenBank mRNA | BC121587.1 | - | 1 - 2 | 132 | 101 | |
| CR760518.2 | - | 1 - 2 | 146 | 101 | ||
More Information
Set Membership: |
|

ifnar2.1
Genomic Map