Assay Details
| Gene Symbol: cdca9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078925.1 | NP_001072393.1 | 6 - 7 | 608 | 105 | |
| GenBank mRNA | BC121590.1 | - | 6 - 7 | 608 | 105 | |
More Information
Set Membership: |
|

cdca9
Genomic Map