Assay Details
| Gene Symbol: alkbh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078926.1 | NP_001072394.1 | 5 - 6 | 576 | 93 | |
| XM_012968616.2 | XP_012824070.1 | 5 - 6 | 737 | 93 | ||
| GenBank mRNA | BC121591.1 | - | 5 - 6 | 576 | 93 | |
More Information
Set Membership: |
|

alkbh1
Genomic Map