Assay Details
| Gene Symbol: ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078928.1 | NP_001072396.1 | 4 - 5 | 257 | 73 | |
| GenBank mRNA | BC121596.1 | - | 4 - 5 | 257 | 73 | |
| BC135869.1 | - | 4 - 5 | 251 | 73 | ||
| CR942734.2 | - | 4 - 5 | 237 | 73 | ||
More Information
Set Membership: |
|

ndufs7
Genomic Map