Assay Details
| Gene Symbol: ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078943.1 | NP_001072411.1 | 1 - 2 | 134 | 72 | |
| XM_012953176.2 | XP_012808630.1 | 2 - 3 | 208 | 72 | ||
| XM_012953177.2 | XP_012808631.1 | 2 - 3 | 208 | 72 | ||
| GenBank mRNA | BC121633.1 | - | 1 - 2 | 134 | 72 | |
| CU024929.1 | - | 2 - 3 | 137 | 72 | ||
More Information
Set Membership: |
|

ecsit
Genomic Map