Assay Details
| Gene Symbol: vopp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078958.1 | NP_001072426.1 | 2 - 3 | 451 | 72 | |
| XM_012964901.2 | XP_012820355.1 | 2 - 3 | 776 | 72 | ||
| XM_018094697.1 | XP_017950186.1 | 2 - 3 | 123 | 72 | ||
| XM_018094698.1 | XP_017950187.1 | 2 - 3 | 123 | 72 | ||
| GenBank mRNA | BC121667.1 | - | 2 - 3 | 451 | 72 | |
More Information
Set Membership: |
|

vopp1
Genomic Map