Assay Details
| Gene Symbol: hspa1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078961.1 | NP_001072429.1 | 1 | 1953 | 74 | |
| GenBank mRNA | BC121673.1 | - | 1 | 1953 | 74 | |
More Information
Set Membership: |
|

hspa1l
Genomic Map