Assay Details
| Gene Symbol: gba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078970.1 | NP_001072438.1 | 4 - 5 | 1269 | 108 | |
| XM_012965261.2 | XP_012820715.1 | 4 - 5 | 1231 | 108 | ||
| XM_018094457.1 | XP_017949946.1 | 4 - 5 | 1257 | 108 | ||
| GenBank mRNA | BC121688.1 | - | 4 - 5 | 1269 | 108 | |
More Information
Set Membership: |
|

gba3
Genomic Map