Assay Details
| Gene Symbol: gba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078970.1 | NP_001072438.1 | 5 - 6 | 1389 | 103 | |
| XM_012965261.2 | XP_012820715.1 | 5 - 6 | 1351 | 103 | ||
| XM_018094457.1 | XP_017949946.1 | 5 - 6 | 1377 | 103 | ||
| GenBank mRNA | BC121688.1 | - | 5 - 6 | 1389 | 103 | |
More Information
Set Membership: |
|

gba3
Genomic Map