Assay Details
| Gene Symbol: igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078972.1 | NP_001072440.1 | 5 - 6 | 902 | 100 | |
| GenBank mRNA | BC121690.1 | - | 5 - 6 | 902 | 100 | |
More Information
Set Membership: |
|

igbp1
Genomic Map