Assay Details
| Gene Symbol: galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078976.1 | NP_001072444.1 | 4 - 5 | 597 | 65 | |
| GenBank mRNA | BC121700.1 | - | 4 - 5 | 597 | 65 | |
More Information
Set Membership: |
|

galnt10
Genomic Map