Assay Details
| Gene Symbol: ca13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078980.1 | NP_001072448.1 | 2 - 3 | 329 | 94 | |
| GenBank mRNA | BC121705.1 | - | 2 - 3 | 329 | 94 | |
More Information
Set Membership: |
|

ca13
Genomic Map