Assay Details
| Gene Symbol: rabl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078983.1 | NP_001072451.1 | 3 - 4 | 294 | 100 | |
| XM_012959242.2 | XP_012814696.1 | 4 - 5 | 406 | 100 | ||
| XM_012959243.2 | XP_012814697.1 | 4 - 5 | 355 | 100 | ||
| GenBank mRNA | BC121708.1 | - | 3 - 4 | 294 | 100 | |
More Information
Set Membership: |
|

rabl2b
Genomic Map