Assay Details
| Gene Symbol: rabl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078983.1 | NP_001072451.1 | 6 - 7 | 583 | 88 | |
| XM_012959242.2 | XP_012814696.1 | 7 - 8 | 695 | 88 | ||
| XM_012959243.2 | XP_012814697.1 | 7 - 8 | 644 | 88 | ||
| GenBank mRNA | BC121708.1 | - | 6 - 7 | 583 | 88 | |
| BC135666.1 | - | 7 - 8 | 624 | 88 | ||
More Information
Set Membership: |
|

rabl2b
Genomic Map