Assay Details
| Gene Symbol: rabl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078983.1 | NP_001072451.1 | 7 - 8 | 666 | 86 | |
| XM_012959242.2 | XP_012814696.1 | 8 - 9 | 778 | 86 | ||
| XM_012959243.2 | XP_012814697.1 | 8 - 9 | 727 | 86 | ||
| GenBank mRNA | BC121708.1 | - | 7 - 8 | 666 | 86 | |
| BC135666.1 | - | 8 - 9 | 707 | 86 | ||
More Information
Set Membership: |
|

rabl2b
Genomic Map