Assay Details
| Gene Symbol: csrnp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078993.1 | NP_001072461.1 | 2 - 3 | 267 | 73 | |
| GenBank mRNA | BC121867.1 | - | 2 - 3 | 267 | 73 | |
More Information
Set Membership: |
|

csrnp1
Genomic Map