Assay Details
| Gene Symbol: fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078995.1 | NP_001072463.1 | 1 - 2 | 856 | 77 | |
| GenBank mRNA | BC121869.1 | - | 1 - 2 | 856 | 77 | |
More Information
Set Membership: |
|

fezf1
Genomic Map