Assay Details
| Gene Symbol: ednrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079008.1 | NP_001072476.1 | 7 - 8 | 1438 | 67 | |
| XM_012957268.2 | XP_012812722.1 | 7 - 8 | 1336 | 67 | ||
| XM_012957269.2 | XP_012812723.1 | 7 - 8 | 1403 | 67 | ||
| XM_012957271.2 | XP_012812725.1 | 7 - 8 | 1127 | 67 | ||
| GenBank mRNA | BC121898.1 | - | 7 - 8 | 1438 | 67 | |
| CR942774.2 | - | 7 - 8 | 1336 | 67 | ||
More Information
Set Membership: |
|

ednrb
Genomic Map