Assay Details
| Gene Symbol: mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079009.1 | NP_001072477.1 | 9 - 10 | 1378 | 71 | |
| GenBank mRNA | BC121899.1 | - | 9 - 10 | 1378 | 71 | |
More Information
Set Membership: |
|

mtfr2
Genomic Map