Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 6 - 7 | 771 | 77 | |
| XM_018097149.1 | XP_017952638.1 | 6 - 7 | 464 | 77 | ||
| GenBank mRNA | BC122975.1 | - | 6 - 7 | 717 | 77 | |
| CT030423.1 | - | 6 - 7 | 726 | 77 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map