Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 9 - 10 | 1734 | 75 | |
| XM_018097149.1 | XP_017952638.1 | 9 - 10 | 1427 | 75 | ||
| GenBank mRNA | BC122975.1 | - | 9 - 10 | 1680 | 75 | |
| CT030423.1 | - | 9 - 10 | 1689 | 75 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map