Assay Details
| Gene Symbol: glyctk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079160.1 | NP_001072628.1 | 3 - 4 | 1079 | 70 | |
| GenBank mRNA | BC123062.1 | - | 3 - 4 | 1079 | 70 | |
| CT030362.1 | - | 4 - 5 | 805 | 70 | ||
More Information
Set Membership: |
|

glyctk
Genomic Map