Assay Details
| Gene Symbol: bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079165.1 | NP_001072633.1 | 10 - 11 | 1572 | 65 | |
| XM_012965653.2 | XP_012821107.1 | 12 - 13 | 1693 | 65 | ||
| XM_012965656.2 | XP_012821110.1 | 12 - 13 | 1750 | 65 | ||
| GenBank mRNA | BC123079.1 | - | 10 - 11 | 1572 | 65 | |
| CU075446.1 | - | 9 - 10 | 1439 | 65 | ||
More Information
Set Membership: |
|

bmpr1b
Genomic Map