Assay Details
| Gene Symbol: tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079180.1 | NP_001072648.1 | 5 - 6 | 800 | 65 | |
| XM_012965704.2 | XP_012821158.1 | 5 - 6 | 1136 | 65 | ||
| XM_012965711.2 | XP_012821165.1 | 5 - 6 | 757 | 65 | ||
| XM_012965717.2 | XP_012821171.1 | 5 - 6 | 743 | 65 | ||
| GenBank mRNA | BC123930.1 | - | 5 - 6 | 800 | 65 | |
More Information
Set Membership: |
|

tfeb
Genomic Map