Assay Details
| Gene Symbol: c1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079184.1 | NP_001072652.1 | 4 - 5 | 452 | 57 | |
| XM_012961365.2 | XP_012816819.1 | 4 - 5 | 484 | 57 | ||
| XM_018093103.1 | XP_017948592.1 | 4 - 5 | 472 | 57 | ||
| GenBank mRNA | BC123935.1 | - | 4 - 5 | 452 | 57 | |
More Information
Set Membership: |
|

c1orf21
Genomic Map