Assay Details
| Gene Symbol: cyp4v2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079199.1 | NP_001072667.1 | 10 - 11 | 1468 | 65 | |
| GenBank mRNA | BC123986.1 | - | 10 - 11 | 1468 | 65 | |
| CU075499.1 | - | 10 - 11 | 1519 | 65 | ||
More Information
Set Membership: |
|

cyp4v2
Genomic Map