Assay Details
| Gene Symbol: cyp4v2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079199.1 | NP_001072667.1 | 4 - 5 | 661 | 84 | |
| GenBank mRNA | BC123986.1 | - | 4 - 5 | 661 | 84 | |
| CU075499.1 | - | 4 - 5 | 712 | 84 | ||
More Information
Set Membership: |
|

cyp4v2
Genomic Map