Assay Details
| Gene Symbol: ednrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079208.1 | NP_001072676.1 | 3 - 4 | 738 | 91 | |
| XM_012968683.2 | XP_012824137.1 | 3 - 4 | 834 | 91 | ||
| XM_012968684.2 | XP_012824138.1 | 3 - 4 | 837 | 91 | ||
| GenBank mRNA | BC124004.1 | - | 3 - 4 | 738 | 91 | |
More Information
Set Membership: |
|

ednrb2
Genomic Map