Assay Details
| Gene Symbol: tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079232.1 | NP_001072700.1 | 4 - 5 | 475 | 67 | |
| XM_012963168.1 | XP_012818622.1 | 4 - 5 | 546 | 67 | ||
| GenBank mRNA | BC124044.1 | - | 4 - 5 | 475 | 67 | |
More Information
Set Membership: |
|

tagap
Genomic Map