Assay Details
| Gene Symbol: ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079234.1 | NP_001072702.1 | 5 - 6 | 751 | 78 | |
| GenBank mRNA | BC124046.1 | - | 5 - 6 | 751 | 78 | |
More Information
Set Membership: |
|

ube2r2
Genomic Map