Assay Details
| Gene Symbol: tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079243.1 | NP_001072711.1 | 10 - 11 | 997 | 74 | |
| XM_012952577.2 | XP_012808031.1 | 10 - 11 | 1464 | 74 | ||
| XM_012952578.2 | XP_012808032.1 | 10 - 11 | 1403 | 74 | ||
| XM_012952579.2 | XP_012808033.1 | 10 - 11 | 1400 | 74 | ||
| XM_012952580.2 | XP_012808034.1 | 9 - 10 | 1365 | 74 | ||
| XM_012952581.2 | XP_012808035.1 | 9 - 10 | 1362 | 74 | ||
| GenBank mRNA | BC124064.1 | - | 10 - 11 | 997 | 74 | |
| BC135831.1 | - | 10 - 11 | 1334 | 74 | ||
More Information
Set Membership: |
|

tlk2
Genomic Map