Assay Details
| Gene Symbol: nos1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079247.1 | NP_001072715.1 | 7 - 8 | 814 | 95 | |
| XM_012961367.2 | XP_012816821.1 | 7 - 8 | 1022 | 95 | ||
| GenBank mRNA | BC125674.1 | - | 7 - 8 | 814 | 95 | |
| CT025320.2 | - | 7 - 8 | 812 | 95 | ||
More Information
Set Membership: |
|

nos1ap
Genomic Map