Assay Details
| Gene Symbol: gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079257.1 | NP_001072725.1 | 3 - 4 | 440 | 65 | |
| XM_012966751.2 | XP_012822205.1 | 4 - 5 | 1189 | 65 | ||
| XM_012966752.2 | XP_012822206.1 | 3 - 4 | 1497 | 65 | ||
| XM_012966753.2 | XP_012822207.1 | 4 - 5 | 1547 | 65 | ||
| GenBank mRNA | BC125693.1 | - | 3 - 4 | 440 | 65 | |
| CT030407.1 | - | 3 - 4 | 423 | 65 | ||
More Information
Set Membership: |
|

gmcl1
Genomic Map