Assay Details
| Gene Symbol: htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079262.2 | NP_001072730.2 | 3 - 4 | 1223 | 78 | |
| GenBank mRNA | BC125705.1 | - | 3 - 4 | 1223 | 78 | |
| CT025323.2 | - | 3 - 4 | 1130 | 78 | ||
| CX740928.1 | - | 399 | 78 | |||
More Information
Set Membership: |
|

htra1
Genomic Map