Assay Details
| Gene Symbol: tyro3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079263.1 | NP_001072731.1 | 12 - 13 | 2014 | 58 | |
| XM_012968690.2 | XP_012824144.1 | 13 - 14 | 2212 | 58 | ||
| GenBank mRNA | BC125706.1 | - | 12 - 13 | 2014 | 58 | |
| CT030338.1 | - | 12 - 13 | 1986 | 58 | ||
More Information
Set Membership: |
|

tyro3
Genomic Map