Assay Details
| Gene Symbol: shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079276.1 | NP_001072744.1 | 5 - 6 | 3558 | 66 | |
| XM_018096333.1 | XP_017951822.1 | 6 - 7 | 3711 | 66 | ||
| XM_018096334.1 | XP_017951823.1 | 7 - 8 | 3777 | 66 | ||
| GenBank mRNA | BC125736.1 | - | 5 - 6 | 3558 | 66 | |
More Information
Set Membership: |
|

shroom4
Genomic Map