Assay Details
| Gene Symbol: prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079284.1 | NP_001072752.1 | 1629 | 56 | ||
| GenBank mRNA | BC125756.1 | - | 1629 | 56 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

prkca| Gene Symbol: prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079284.1 | NP_001072752.1 | 1629 | 56 | ||
| GenBank mRNA | BC125756.1 | - | 1629 | 56 | ||
Set Membership: |
|