Assay Details
| Gene Symbol: mrtfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079286.1 | NP_001072754.1 | 13 - 14 | 2346 | 73 | |
| XM_012961404.2 | XP_012816858.1 | 13 - 14 | 2451 | 73 | ||
| XM_012961405.2 | XP_012816859.1 | 13 - 14 | 2461 | 73 | ||
| XM_012961406.2 | XP_012816860.1 | 11 - 12 | 2384 | 73 | ||
| XM_012961407.2 | XP_012816861.1 | 14 - 15 | 2530 | 73 | ||
| XM_018093106.1 | XP_017948595.1 | 12 - 13 | 2333 | 73 | ||
| XM_018093107.1 | XP_017948596.1 | 11 - 12 | 1997 | 73 | ||
| GenBank mRNA | BC125766.1 | - | 13 - 14 | 2346 | 73 | |
More Information
Set Membership: |
|

mrtfa
Genomic Map