Assay Details
| Gene Symbol: mrtfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079286.1 | NP_001072754.1 | 9 - 10 | 1029 | 64 | |
| XM_012961404.2 | XP_012816858.1 | 9 - 10 | 1134 | 64 | ||
| XM_012961405.2 | XP_012816859.1 | 9 - 10 | 1144 | 64 | ||
| XM_012961406.2 | XP_012816860.1 | 7 - 8 | 1067 | 64 | ||
| XM_012961407.2 | XP_012816861.1 | 10 - 11 | 1213 | 64 | ||
| XM_018093107.1 | XP_017948596.1 | 7 - 8 | 680 | 64 | ||
| GenBank mRNA | BC125766.1 | - | 9 - 10 | 1029 | 64 | |
More Information
Set Membership: |
|

mrtfa
Genomic Map