Assay Details
| Gene Symbol: msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079341.1 | NP_001072809.1 | 5 - 6 | 871 | 71 | |
| GenBank mRNA | BC121456.1 | - | 5 - 6 | 871 | 71 | |
More Information
Set Membership: |
|

msmo1
Genomic Map