Assay Details
| Gene Symbol: kmt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079347.1 | NP_001072815.1 | 5 - 6 | 691 | 81 | |
| XM_018094658.1 | XP_017950147.1 | 4 - 5 | 639 | 81 | ||
| GenBank mRNA | BC121601.1 | - | 5 - 6 | 691 | 81 | |
More Information
Set Membership: |
|

kmt5a
Genomic Map