Assay Details
| Gene Symbol: adad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079364.1 | NP_001072832.1 | 4 - 5 | 603 | 74 | |
| GenBank mRNA | BC121902.1 | - | 4 - 5 | 603 | 74 | |
More Information
Set Membership: |
|

adad2
Genomic Map