Assay Details
| Gene Symbol: mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079365.1 | NP_001072833.1 | 6 - 7 | 1083 | 59 | |
| GenBank mRNA | BC121909.1 | - | 6 - 7 | 1083 | 59 | |
| BC135638.1 | - | 6 - 7 | 1068 | 59 | ||
More Information
Set Membership: |
|

mtmr9
Genomic Map