Assay Details
| Gene Symbol: cybc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079369.1 | NP_001072837.1 | 2 - 3 | 131 | 101 | |
| GenBank mRNA | BC121946.1 | - | 2 - 3 | 131 | 101 | |
| CT025438.2 | - | 2 - 3 | 140 | 101 | ||
More Information
Set Membership: |
|

cybc1
Genomic Map