Assay Details
| Gene Symbol: cybc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079369.1 | NP_001072837.1 | 4 - 5 | 246 | 104 | |
| GenBank mRNA | BC121946.1 | - | 4 - 5 | 246 | 104 | |
| CT025438.2 | - | 4 - 5 | 255 | 104 | ||
More Information
Set Membership: |
|

cybc1
Genomic Map